missing translation for 'onlineSavingsMsg'
Learn More

Dalfopristin, MedChemExpress

Product Code. p-200100983
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16440748

Brand: Medchem Express HYA0241

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dalfopristin is a semi-synthetic streptogramin antibiotic. Quinupristin/Dalfopristin (Q/D) is a valuable alternative antibiotic to vancomycin for the treatment of multi-drug resistant Enterococcus faecium infections.

Dalfopristin is a semi-synthetic streptogramin antibiotic. Quinupristin/Dalfopristin (Q/D) is a valuable alternative antibiotic to vancomycin for the treatment of multi-drug resistant Enterococcus faecium infections.

Specifications

Chemical Name or Material Dalfopristin
Physical Form Solid
CAS 112362-50-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C34H50N4O9S
For Use With (Application) COVID-19-immunoregulation
Synonym RP54476
Solubility Information DMSO : 125 mg/mL (180.94 mM; Need ultrasonic)
SMILES O=S(CCN(CC)CC)([C@H]1[C@@](C(O[C@H]([C@@H](/C=C/C(NC/C=C\C(C)=C\2)=O)C)C(C)C)=O)([H])N(C(C3=COC(CC(C[C@@H]2O)=O)=N3)=O)CC1)=O
Molecular Weight (g/mol) 690.85
Formula Weight 690.85
Percent Purity 98.34%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.