missing translation for 'onlineSavingsMsg'
Learn More

Dalfampridine, TRC

Product Code. 30360574
Change view
Click to view available options
Quantity:
1 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30360574 250 g
30350196 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30360574 Supplier Toronto Research Chemicals Supplier No. TRCD113500250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-24-5
Molecular Formula C5 H6 N2
Molecular Weight (g/mol) 94.11
Synonym Pyridine, 4-amino- (8CI), 4-Aminopyridine, 4-Pyridinylamine, 4-Pyridylamine, 4AP, Ampyra, Avitrol, Dalfampridine, EL 970, Fampridine, NSC 15041, Neurelan, Phillips 1861, Pymadin, VMI 10-3, p-Aminopyridine, γ-Aminopyridine
IUPAC Name pyridin-4-amine
SMILES Nc1ccncc1

Specifications

Quantity 250 g
InChI Formula InChI=1S/C5H6N2/c6-5-1-3-7-4-2-5/h1-4H,(H2,6,7)
Formula Weight 94.0531
Chemical Name or Material 4-Aminopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.