missing translation for 'onlineSavingsMsg'
Learn More

Dalfampridine, Mikromol™

Product Code. 18083750
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18083750 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18083750 Supplier Mikromol Supplier No. MM3462.00
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dalfampridine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminopyridine
CAS 504-24-5
Analyte or Component Names 4-Aminopyridine
Shipping Condition Cold 2°C to 8°C
Molecular Formula C5 H6 N2
Certifications/Compliance ISO 17025
Synonym 4-Pyridinamine, Pyridine, 4-amino- (8CI), 4-Aminopyridine, 4-Pyridinylamine, 4-Pyridylamine, 4AP, Ampyra, Avitrol, Dalfampridine, EL 970, Fampridine, NSC 15041, Neurelan, Phillips 1861, VMI 10-3, p-Aminopyridine, gamma-Aminopyridine
InChI Formula InChI=1S/C5H6N2/c6-5-1-3-7-4-2-5/h1-4H,(H2,6,7)
SMILES Nc1ccncc1
IUPAC Name pyridin-4-amine
Molecular Weight (g/mol) 94.11
Quantity 100 mg
Formula Weight 94.0531 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.