missing translation for 'onlineSavingsMsg'
Få mere at vide

Dalapon-methyl-D3, TRC

Artikelnummer. 30384313
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
25 mg
100 mg
250 mg
Pakningsstørrelse:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30384313 100 mg 100mg
30380068 250 mg 250mg
30381921 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30384313 Leverandør Toronto Research Chemicals Leverandørnr. TRCD458131100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Dalapon-methyl-D3
Molecular Formula C4 D3 H3 Cl2 O2
Quantity 100 mg
Synonym Methyl 2,2-Dichloropropanoate-D3, Methyl 2,2-dichloropropionate-D3, α,α-Dichloropropionic Acid Methyl ester-D3
InChI Formula InChI=1S/C4H6Cl2O2/c1-4(5,6)3(7)8-2/h1-2H3/i2D3
SMILES [2H]C([2H])([2H])OC(=O)C(C)(Cl)Cl
IUPAC Name trideuteriomethyl 2,2-dichloropropanoate
Molecular Weight (g/mol) 160.014
Formula Weight 158.993
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.