missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DAK Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA527842

Product Code. 13431987

  • £439.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: HepG2, Raji. Predicted reactivity: Mouse (82%), Rat (83%), Chimpanzee (100%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of the family of dihydroxyacetone kinases, which have a protein structure distinct from other kinases. The product of this gene phosphorylates dihydroxyacetone, and also catalyzes the formation of riboflavin 4',5'-phosphate (aka cyclin FMN) from FAD. Several alternatively spliced transcript variants have been identified, but the full-length nature of only one has been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DAK
Polyclonal
Unconjugated
TKFC
ATP-dependent dihydroxyacetone kinase; BC021917; bifunctional ATP-dependent dihydroxyacetone kinase/FAD-AMP lyase (cyclizing); Dak; DHA kinase; Dha kinase/FMN cyclase; dihydroxyacetone kinase 2 homolog; dihydroxyacetone kinase 2 homolog (S. cerevisiae); DKFZP586B1621; DKFZP586B1621 protein; FAD-AMP lyase (cyclic FMN forming); FAD-AMP lyase (cyclizing); FAD-AMP lyase cyclic FMN forming; FAD-AMP lyase cyclizing; FMN cyclase; Glycerone kinase; MGC5621; NET45; RGD1311026; testis tissue sperm-binding protein Li 84P; TKFC; Triokinase; triokinase and FMN cyclase; triokinase, FMN cyclase; triokinase/FMN cyclase; Triose kinase; zgc:153296
Rabbit
Antigen affinity chromatography
RUO
26007
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.76 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q3LXA3
TKFC
Recombinant protein encompassing a sequence within the center region of human DAK. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.