missing translation for 'onlineSavingsMsg'
Learn More

DAK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00 - £513.00

Specifications

Antigen DAK
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18688275
View Documents
Novus Biologicals
NBP2-48894-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18664536
View Documents
Novus Biologicals
NBP2-48894
0.1 mL
£513.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DAK Polyclonal antibody specifically detects DAK in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

DAK
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Lipid and Metabolism
PBS (pH 7.2), 40% Glycerol
26007
IgG
Immunogen affinity purified
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Purified
RUO
Human
ATP-dependent dihydroxyacetone kinase, bifunctional ATP-dependent dihydroxyacetone kinase/FAD-AMP lyase (cyclizing), DHA kinase, Dha kinase/FMN cyclase, dihydroxyacetone kinase 2 homolog (S. cerevisiae), dihydroxyacetone kinase 2 homolog (yeast), DKFZp586B1621, FAD-AMP lyase cyclic FMN forming, FAD-AMP lyase cyclizing, glycerone kinase, MGC5621, NET45
This antibody was developed against a recombinant protein corresponding to amino acids: PGDRTMLDSLWAAGQELQAWKSPGADLLQVLTKAVKSAEAAAEATKNMEAGAGRASYISSARLEQPDPGAVAAAAILRAIL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.