missing translation for 'onlineSavingsMsg'
Learn More

Daidzein 7-Beta-D-Glucuronide 4’-Sulfate Disodium Salt, TRC

Product Code. 30006261
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006261 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30006261 Supplier Toronto Research Chemicals Supplier No. TRCD1035251MG
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1041134-19-3
Molecular Formula C21 H16 Na2 O13 S
Quantity 1 mg
InChI Formula InChI=1S/C21H18O13S.2Na/c22-15-12-6-5-11(32-21-18(25)16(23)17(24)19(33-21)20(26)27)7-14(12)31-8-13(15)9-1-3-10(4-2-9)34-35(28,29)30;;/h1-8,16-19,21,23-25H,(H,26,27)(H,28,29,30);;/q;2*+1/p-2/t16-,17-,18+,19-,21+;;/m0../s1
SMILES [Na+].[Na+].O[C@H]1[C@H](Oc2ccc3C(=O)C(=COc3c2)c4ccc(OS(=O)(=O)[O-])cc4)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)[O-]
IUPAC Name disodium;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-oxo-3-(4-sulfonatooxyphenyl)chromen-7-yl]oxyoxane-2-carboxylate
Molecular Weight (g/mol) 554.39
Chemical Name or Material Daidzein 7-beta-D-Glucuronide 4'-sulfate disodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.