missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Daidzein

Product Code. 16402346
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402346 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402346 Supplier MedChemExpress Supplier No. HYN0019
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Daidzein is a soy isoflavone, which acts as a PPAR activator.

Specifications

Content And Storage 4°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
Product Type Natural Products
Purity or Quality Grade 98.0%
Quantity 500 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 50 mg/mL (196.66 mM) ∣H2O : < 0.1 mg/mL (insoluble)
Target PPAR∣Endogenous Metabolite
CAS 486-66-8
Molecular Weight (g/mol) 254.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.