missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAGLB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567047
This item is not returnable.
View return policy
Description
Immunogen sequence: LVALDHRKESV VVAVRGTMSL QDVLTDLSAE SEVLDVECEV QDRLAHKGIS QAARYVYQRL INDGILSQAF SIA Highest antigen sequence identity to the following orthologs - mouse 78%, rat 80%.
Lipase that catalyzes the hydrolysis of arachidonic acid (AA)-esterified diacylglycerols (DAGs) to produce the principal endocannabinoid, 2-arachidonoylglycerol (2-AG) which can be further cleaved by downstream enzymes to release arachidonic acid (AA) for cyclooxygenase (COX)-mediated eicosanoid production (PubMed:14610053). Preferentially hydrolyzes DAGs at the sn-1 position in a calcium-dependent manner and has negligible activity against other lipids including monoacylglycerols and phospholipids (PubMed:14610053). Plays a key role in the regulation of 2-AG and AA pools utilized by COX1/2 to generate lipid mediators of macrophage and microglia inflammatory responses. Functions also as a polyunsaturated fatty acids-specific triacylglycerol lipase in macrophages. Plays an important role to support the metabolic and signaling demands of macrophages. [UniProt].
Specifications
| DAGLB | |
| Polyclonal | |
| Unconjugated | |
| DAGLB | |
| DAGLB; DAGLBETA; DGL-beta; diacylglycerol lipase beta; diacylglycerol lipase, beta; E330036I19Rik; KCCR13L; RGD1310193; sn1-specific diacylglycerol lipase beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 221955 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NCG7 | |
| DAGLB | |
| Recombinant Human DAGLB. Recombinant protein control fragment (Product #RP-105476). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu