missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAGLA Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-20905
This item is not returnable.
View return policy
Description
DAGLA Polyclonal specifically detects DAGLA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| DAGLA | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| C11orf11, DAGL(ALPHA), DAGLALPHA, DGL-alpha, diacylglycerol lipase, alpha, EC 3.1.1.-, KIAA0659, Neural stem cell-derived dendrite regulator, NSDDRchromosome 11 open reading frame 11, sn1-specific diacylglycerol lipase alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected from sequence similarity: Mouse and Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:128000 | |
| NP_006124.1 | |
| DAGLA | |
| Peptide with sequence C-PAKQDELVISAR corresponding to C-Terminus according to NP_006124.1. | |
| 0.1 mg | |
| Cellular Markers | |
| 747 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction