missing translation for 'onlineSavingsMsg'
Learn More

Dafadine-A, MedChemExpress

Product Code. p-200100625
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16439657

Brand: Medchem Express HY16670

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dafadine-A, an analog of dafadine, is a novel inhibitor of DAF-9 cytochrome P450 in the nematode Caenorhabditis elegans; also inhibits the mammalian ortholog of DAF-9(CYP27A1).

Dafadine-A, an analog of dafadine, is a novel inhibitor of DAF-9 cytochrome P450 in the nematode Caenorhabditis elegans

Specifications

Chemical Name or Material Dafadine-A
CAS 1065506-69-5
Color Brownish Yellow
Purity Grade Notes Research
Molecular Formula C23H25N3O3
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (127.73 mM; Need ultrasonic)
SMILES O=C(N1CCC(C2=CC=NC=C2)CC1)C3=NOC(COC4=C(C)C=CC=C4C)=C3
Molecular Weight (g/mol) 391.46
Formula Weight 391.46
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.