missing translation for 'onlineSavingsMsg'
Learn More

Dafadine-A, MedChemExpress

Product Code. 16439657
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439657 5 mg 5mg
16437118 10 mg 10mg
16413157 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16439657 Supplier Medchem Express Supplier No. HY16670

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dafadine-A, an analog of dafadine, is a novel inhibitor of DAF-9 cytochrome P450 in the nematode Caenorhabditis elegans; also inhibits the mammalian ortholog of DAF-9(CYP27A1).

Dafadine-A, an analog of dafadine, is a novel inhibitor of DAF-9 cytochrome P450 in the nematode Caenorhabditis elegans

Specifications

Chemical Name or Material Dafadine-A
CAS 1065506-69-5
Color Brownish Yellow
Purity Grade Notes Research
Molecular Formula C23H25N3O3
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (127.73 mM; Need ultrasonic)
SMILES O=C(N1CCC(C2=CC=NC=C2)CC1)C3=NOC(COC4=C(C)C=CC=C4C)=C3
Molecular Weight (g/mol) 391.46
Formula Weight 391.46
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.