missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520062
This item is not returnable.
View return policy
Description
Despite its predicted molecular weight, DAD1 often migrates at ~22kDa in SDS-PAGE. A suggested positive control is HepG2 cell lysate. PA5-20062 can be used with blocking peptide PEP-0181.
Defender of cell death 1 (DAD1) was initially discovered in BHK21 cells as a negative regulator of programmed cell death, a process important for normal organism development and tissue homeostasis. DAD1 was later shown to be a subunit of the mammalian oligosaccharyltransferase complex and is required for its function and structural integrity. Mice lacking DAD1 express abnormal N-linked glycoproteins and undergo increased apoptotic-associated embryonic death. Furthermore, overexpression of DAD1 mRNA is seen in some human hepatocellular carcinomas, indicating it may also play a role in carcinogenesis. It should be noted that DAD1 is not related to the inhibitor of apoptosis proteins (IAP) family and does not contain any baculoviral IAP repeat (BIR) domains.
Specifications
| DAD1 | |
| Polyclonal | |
| Unconjugated | |
| DAD1 | |
| AI323713; Dad1; DAD-1; Defender against cell death 1; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit DAD1; Oligosaccharyl transferase subunit DAD1; oligosaccharyltransferase 2 homolog; oligosaccharyltransferase subunit 2 (non-catalytic); OST2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13135, 1603 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P61803, P61804 | |
| DAD1 | |
| A peptide corresponding to 14 amino acids near the C-terminus of human DAD1. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction