missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAD1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
£436.00
Specifications
| Antigen | DAD1 |
|---|---|
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Form | Purified |
Description
DAD1 Polyclonal antibody specifically detects DAD1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| DAD1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DAD-1, defender against cell death 1EC 2.4.1.119, dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit DAD1, Oligosaccharyl transferase subunit DAD1, oligosaccharyltransferase 2 homolog, OST2 | |
| Antibody was raised against a peptide corresponding to 14 amino acids near the C-terminus of human DAD1. The immunogen is located within the last 50 amino acids of DAD1. Amino Acid Squence: CLRIQINPQNKAD | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 500 SE | |
| Rabbit | |
| Apoptosis, Cancer | |
| 50 mM Sodium Borate | |
| 1603 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title