missing translation for 'onlineSavingsMsg'
Learn More

DAD1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18425913 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425913 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425913 Supplier Novus Biologicals Supplier No. NBP177197MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DAD1 Polyclonal antibody specifically detects DAD1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAD1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DAD-1, defender against cell death 1EC 2.4.1.119, dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit DAD1, Oligosaccharyl transferase subunit DAD1, oligosaccharyltransferase 2 homolog, OST2
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the C-terminus of human DAD1. The immunogen is located within the last 50 amino acids of DAD1. Amino Acid Squence: CLRIQINPQNKAD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1603
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.