missing translation for 'onlineSavingsMsg'
Learn More

DAD1 Antibody (2B4-C8), Novus Biologicals™

Product Code. 18381238 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381238 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381238 Supplier Novus Biologicals Supplier No. H00001603M0350ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DAD1 Monoclonal antibody specifically detects DAD1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAD1
Applications ELISA
Classification Monoclonal
Clone 2B4-C8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH07403
Gene Alias DAD-1, defender against cell death 1EC 2.4.1.119, dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit DAD1, Oligosaccharyl transferase subunit DAD1, oligosaccharyltransferase 2 homolog, OST2
Host Species Mouse
Immunogen DAD1 (AAH07403, 1 a.a. ~ 113 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSASVVSVISRFLEEYLSSTPQRLKLLDAYLLYILLTGALQFGYCLLVGTFPFNSFLSGFISCVGSFILAVCLRIQINPQNKADFQGISPERAFADFLFASTILHLVVMNFVG
Purification Method IgG purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1603
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.