missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DACT3/Dapper 3 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76340CL1
This item is not returnable.
View return policy
Description
DACT3/Dapper 3 Polyclonal antibody specifically detects DACT3/Dapper 3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| DACT3/Dapper 3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| Antagonist of beta-catenin Dapper homolog 3, arginine rich region 1, Arginine-rich region 1 protein, dapper homolog 3, dapper, antagonist of beta-catenin, homolog 3 (Xenopus laevis), MGC15476, RRR1 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human Dact3. The immunogen is located within amino acids 370 - 420 of Dact3. Amino Acid Squence: SVEQSPPRERPRAAGRR | |
| 0.1 mL | |
| Neuroscience, Stem Cells | |
| 147906 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction