missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DACT3/Dapper 3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | DACT3/Dapper 3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
DACT3/Dapper 3 Polyclonal specifically detects DACT3/Dapper 3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| DACT3/Dapper 3 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q96B18 | |
| 147906 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human Dact3. The immunogen is located within amino acids 370 - 420 of Dact3. Amino Acid Squence: SVEQSPPRERPRAAGRR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antagonist of beta-catenin Dapper homolog 3, arginine rich region 1, Arginine-rich region 1 protein, dapper homolog 3, dapper, antagonist of beta-catenin, homolog 3 (Xenopus laevis), MGC15476, RRR1 | |
| DACT3 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title