missing translation for 'onlineSavingsMsg'
Learn More

DACT3/Dapper 3 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18419306 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419306 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419306 Supplier Novus Biologicals Supplier No. NBP176340AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DACT3/Dapper 3 Polyclonal antibody specifically detects DACT3/Dapper 3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DACT3/Dapper 3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias Antagonist of beta-catenin Dapper homolog 3, arginine rich region 1, Arginine-rich region 1 protein, dapper homolog 3, dapper, antagonist of beta-catenin, homolog 3 (Xenopus laevis), MGC15476, RRR1
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human Dact3. The immunogen is located within amino acids 370 - 420 of Dact3. Amino Acid Squence: SVEQSPPRERPRAAGRR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 147906
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.