missing translation for 'onlineSavingsMsg'
Learn More

DACT3/Dapper 3 Antibody (2A5), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18423414 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423414 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423414 Supplier Novus Biologicals Supplier No. NBP119052AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DACT3/Dapper 3 Monoclonal antibody specifically detects DACT3/Dapper 3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DACT3/Dapper 3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2A5
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Accession No. NP_659493
Gene Alias Antagonist of beta-catenin Dapper homolog 3, arginine rich region 1, Arginine-rich region 1 protein, dapper homolog 3, dapper, antagonist of beta-catenin, homolog 3 (Xenopus laevis), MGC15476, RRR1
Host Species Mouse
Immunogen Recombinant human DACT3/Dapper 3 (89-202aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 147906
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.