missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DACT2 Polyclonal Antibody

Description
A suggested positive control is SK-N-SH cell lysate. PA5-20629 can be used with blocking peptide PEP-0749.
The Wnt signaling cascade is a conserved process in multicellular animals that plays important roles during development and can contribute to cancer and other diseases. Many members of this pathway are also expressed in the postnatal tissues such as brain. One such protein is Dact2, a member of the Dact protein family that was initially identified through binding to Disheveled (Dvl), a cytoplasmic protein essential to Wnt signaling. Dact2 is most prominent during the development of the thymus kidneys, and salivary gland. Dact2 is thought to play a role distinct from that of Dact1 with Dact2 having a greater impact on a beta-catenin-independent process termed planar cell polarity/convergent-extension signaling. Furthermore, Dact2 but not Dact1 can inhibit Nodal signaling by promoting the endocytic degradation of TGF-beta receptors. At least two isoforms of Dact2 are known to exist.
Specifications
Specifications
| Antigen | DACT2 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | DACT2 |
| Gene Accession No. | Q5SW24, Q7TN08 |
| Gene Alias | 2900084M21Rik; A630024E20; bA503C24.7; C6orf116; Dact2; Dapper antagonist of catenin 2; dapper homolog 2; dapper homolog 2, antagonist of beta-catenin; dapper homolog 2, antagonist of beta-catenin (xenopus); dapper, antagonist of beta-catenin, homolog 2; DAPPER2; dishevelled binding antagonist of beta catenin 2; dishevelled-binding antagonist of beta-catenin 2; Dpr2; Frd2; mDpr2; PP13671; RP11-503C24.7 |
| Gene Symbols | DACT2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?