missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dact2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00
Specifications
| Antigen | Dact2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Dact2 Polyclonal specifically detects Dact2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| Dact2 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_999627 | |
| 168002 | |
| Antibody was raised against a 12 amino acid synthetic peptide from near the amino terminus of human DACT2. The immunogen is located within amino acids 150 - 200 of Dact2. Amino Acid Squence: GDWRPRSVDETT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| bA503C24.7, C6orf116, chromosome 6 open reading frame 116, dapper homolog 2, dapper homolog 2, antagonist of beta-catenin, dapper homolog 2, antagonist of beta-catenin (xenopus), dapper, antagonist of beta-catenin, homolog 2 (Xenopus laevis), DAPPER2, DPR2, FLJ31232, MGC133141, MGC133142, RP11-503C24.7 | |
| DACT2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title