missing translation for 'onlineSavingsMsg'
Learn More

Dact1 Antibody (OTI10F12) [Alexa Fluor™ 647], Novus Biologicals™

Product Code. 30688880 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688880 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688880 Supplier Novus Biologicals Supplier No. NBP344718AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dact1 Monoclonal antibody specifically detects Dact1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dact1
Applications Western Blot
Classification Monoclonal
Clone OTI10F12
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias DAPPER, dapper, antagonist of beta-catenin, homolog 1 (Xenopus laevis), DAPPER1dapper homolog 1, antagonist of beta-catenin (xenopus), DPR1dapper homolog 1, antagonist of beta-catenin, FRODOdapper homolog 1, HDPR1, Hepatocellular carcinoma novel gene 3 protein, heptacellular carcinoma novel gene 3, HNG3, THYEX3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 289-555 of human DACT1(NP_057735) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51339
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.