missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Dacomitinib
GREENER_CHOICE

Product Code. 18719776
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18719776 10 mg 10mg
18735966 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18719776 Supplier Tocris Bioscience™ Supplier No. 6231/10
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent irreversible pan ErbB inhibitor

Dacomitinib is a potent irreversible pan ErbB inhibitor (IC50 values are 6.0, 45.7 and 73.7 nM for EGFR, ERBB2 and ERBB4, respectively). Inhibits mutant and wild type forms of ErbB, including EGFR T780M, and shows activity in NSCLC cell lines sensitive and resistant to Iressa (Cat. No. 3000). Inhibits tumor growth. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-methoxy-6-quinazolinyl]-4-(1-piperidinyl)-2-butenamide
CAS 1110813-31-4
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C24H25ClFN5O2
Formula Weight Observed MW: 469.94
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.