Learn More
Invitrogen™ Daclizumab Chimeric Recombinant Rabbit Monoclonal Antibody (Daclizumab)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547834
Description
Specificity: Binds to p55 chain of IL2R on human T cells, Tac antigen, which is expressed on T cells activated by mitogens, soluble antigens, and alloantigens but not on resting T cells, thymocytes, B cells, monocytes, activated B cells, leukemic T cell blasts from patients with acute lymphoblastic leukemia, and long-term T cell lines.
Humanized IgG1 Mab that binds to the human interleukin-2 receptor (anti-Tac or anti-CD25). Daclizumab is a composite of human (90%) and murine (10%) antibody sequences. The human sequences were derived from the constant domains of human IgG1 and the variable framework regions of the Eu myeloma antibody. The murine sequences were derived from the complementarity-determining regions of a murine anti-Tac antibody. On 22 April 2008, Roche Registration Limited chose to voluntarily withdraw the marketing authorization for their product Zenapax (daclizumab), as indicated for the prophylaxis of acute organ rejection in de novo allogeneic renal transplantation and used concomitantly with an immunosuppressive regimen like cyclosporine and corticosteroids in patients who are not immunized, for commercial reasons and confirmed that this decision was not related to any safety concerns associated with the use of Zenapax (daclizumab) 2. Regardless of the withdrawal of Zenapax, Biogen and Abbvie's Zinbryta (daclizumab), as indicated for the treatment of adult patients with relapsing forms of multiple sclerosis, was approved for use by the FDA in 2016.
Specifications
| Daclizumab Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P01911, P01920, P04440 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3115, 3119, 3123 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Inhibition Assays | |
| Daclizumab | |
| Unconjugated | |
| HLA-DQB1 | |
| BIIB019; DAC HYP | |
| Cultured T-cells derived from a patient with Mycosis fungoides. | |
| 200 μg | |
| Primary | |
| Human, Rhesus Macaque, Cynomolgus Monkey | |
| Antibody | |
| IgG κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.