missing translation for 'onlineSavingsMsg'
Learn More

Daclatasvir, MedChemExpress

Product Code. 16434996
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434996

Brand: Medchem Express HY10466

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Daclatasvir (BMS-790052) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir is also a organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.

Daclatasvir (BMS-790052) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir is also a organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.

Specifications

Chemical Name or Material Daclatasvir
Physical Form Solid
CAS 1009119-64-5
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C40H50N8O6
For Use With (Application) COVID-19-anti-virus
Synonym BMS-790052 EBP 883
Solubility Information DMSO : ≥ 40 mg/mL (54.14 mM)
SMILES O=C([C@@H](NC(OC)=O)C(C)C)N1CCC[C@H]1C2=NC=C(N2)C3=CC=C(C=C3)C4=CC=C(C5=CN=C([C@@H]6CCCN6C([C@@H](NC(OC)=O)C(C)C)=O)N5)C=C4
Molecular Weight (g/mol) 738.88
Formula Weight 738.88
Percent Purity 98.19%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.