missing translation for 'onlineSavingsMsg'
Learn More

Daclatasvir, MedChemExpress

Product Code. 16483988
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483988 10 mg 10mg
16471576 500 mg 500mg
16470986 200 mg 200mg
16434996 100 mg 100mg
16430347 50 mg 50mg
16406707 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16483988 Supplier Medchem Express Supplier No. HY10466
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Daclatasvir (BMS-790052) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir is also a organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.

Daclatasvir (BMS-790052) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir is also a organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.

Specifications

Chemical Name or Material Daclatasvir
Physical Form Solid
CAS 1009119-64-5
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C40H50N8O6
For Use With (Application) COVID-19-anti-virus
Synonym BMS-790052 EBP 883
Solubility Information DMSO : ≥ 40 mg/mL (54.14 mM)
SMILES O=C([C@@H](NC(OC)=O)C(C)C)N1CCC[C@H]1C2=NC=C(N2)C3=CC=C(C=C3)C4=CC=C(C5=CN=C([C@@H]6CCCN6C([C@@H](NC(OC)=O)C(C)C)=O)N5)C=C4
Molecular Weight (g/mol) 738.88
Formula Weight 738.88
Percent Purity 98.19%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.