missing translation for 'onlineSavingsMsg'
Learn More

Dacarbazine, MedChemExpress

Product Code. 16402049
Encompass_Preferred
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
This item is not returnable. View return policy

Product Code. 16402049

Brand: Medchem Express HYB0078

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dacarbazine is a cell cycle nonspecific antineoplastic alkylating agent. Dacarbazine inhibits T and B lymphoblastic response, with IC50 values of 50 and 10 μg/ml, respectively. Dacarbazine can be used for the research of metastatic malignant melanoma.

Dacarbazine is a cell cycle nonspecific antineoplastic alkylating agent. Dacarbazine inhibits T and B lymphoblastic response, with IC50 values of 50 and 10 μg/ml, respectively. Dacarbazine can be used for the research of metastatic malignant melanoma.

Specifications

Chemical Name or Material Dacarbazine
Physical Form Solid
CAS 3-4-4342
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C6H10N6O
For Use With (Application) Cancer-programmed cell death
Synonym Imidazole Carboxamide
Solubility Information DMSO : 5 mg/mL (27.45 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 1 mg/mL (5.49 mM; Need ultrasonic and warming)
SMILES O=C(C1=C(/N=N/N(C)C)NC=N1)N
Molecular Weight (g/mol) 182.18
Formula Weight 182.18
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.