missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dacarbazine, Fisher BioReagents
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass
Unit Size:
250mg
Description
Dacarbazine is an alkylating agent used in cancer research. Antineoplastic.
Specifications
Specifications
| Melting Point | 202°C |
| Color | White |
| Packaging | Amber Glass |
| Quantity | 250 mg |
| Merck Index | 15, 2795 |
| Formula Weight | 182.2 |
| Physical Form | Powder or Solid |
| Chemical Name or Material | Dacarbazine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction