missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dacarbazine, Fisher BioReagents
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass
Description
Dacarbazine is an alkylating agent used in cancer research. Antineoplastic.
Specifications
Melting Point | 202°C |
Color | White |
Packaging | Amber Glass |
Quantity | 250 mg |
Merck Index | 15, 2795 |
Formula Weight | 182.2 |
Physical Form | Powder or Solid |
Chemical Name or Material | Dacarbazine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'