missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dabsyl Chloride, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Dabsyl Chloride |
| CAS | 56512-49-3 |
| Molecular Formula | C14 H14 Cl N3 O2 S |
| Quantity | 5 g |
| Synonym | 4-[[4-(Dimethylamino)phenyl]azo]benzenesulfonyl Chloride, 4-Dimethylaminoazobenzene-4'-sulfonyl Chloride, 4'-(Dimethylamino)azobenzene-4-sulfonic Acid Chloride, DABS-Cl |
| InChI Formula | InChI=1S/C14H14ClN3O2S/c1-18(2)13-7-3-11(4-8-13)16-17-12-5-9-14(10-6-12)21(15,19)20/h3-10H,1-2H3/b17-16+ |
| SMILES | CN(C)c1ccc(cc1)N=Nc2ccc(cc2)S(=O)(=O)Cl |
| IUPAC Name | 4-[(E)-[4-(dimethylamino)phenyl]azo]benzenesulfonyl chloride |
| Molecular Weight (g/mol) | 323.8 |
| Formula Weight | 323.0495 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction