missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Dabrafenib mesylate

Product Code. 18756726
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18756726 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18756726 Supplier R&D Systems™ Supplier No. 7223/50
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective B-Raf, CDK16 and NEK9 inhibitor; anticancer; also targets human proteins in the SARS-CoV-2 interactome

Dabrafenib mesylate is a potent and selective Raf kinase inhibitor (IC50 values are 0.8, 3.2 and 5.0 nM for B-RafV600E, wild-type B-Raf and c-Raf, respectively). Exhibits selectivity for B-Raf over a panel of 270 other kinases. Arrests cell cycle in the G1 phase and inhibits growth of B-RafV600E mutant melanoma and colon cancer cells in vitro and in vivo. Orally bioavailable. Also inhibits NEK9 (IC50 <10 nM) and CDK16. Identified as targeting human host proteins that interact with SARS-CoV-2.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-[5-(2-Aminopyrimidin-4-yl)-2-tert-butyl-1,3-thiazol-4-yl]-2-fluorophenyl]-2,6-difluorobenzenesulfonamide methanesulfonate
CAS 1195768-06-9
Quantity 50 mg
Product Line Tocris
Target Raf Kinase Inhibitors
Molecular Formula C23H20F3N5O2S2.CH3SO3H
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.