missing translation for 'onlineSavingsMsg'
Learn More

Dabrafenib Mesylate, MedChemExpress

Product Code. 16492257
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492257 50 mg 50mg
16478727 5 mg 5mg
16461746 500 mg 500mg
16456516 100 mg 100mg
16426128 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16492257 Supplier Medchem Express Supplier No. HY14660A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dabrafenib Mesylate is a potent and selective Raf kinase inhibitor with IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf, respectively.

Dabrafenib Mesylate is a potent and selective Raf kinase inhibitor with IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf, respectively.

Specifications

Chemical Name or Material Dabrafenib Mesylate
CAS 1195768-06-9
Color White
Purity Grade Notes Research
Molecular Formula C24H24F3N5O5S3
Quantity 50 mg
Synonym GSK2118436 Mesylate GSK 2118436B
Solubility Information DMSO : ≥ 36 mg/mL (58.47 mM)
SMILES CC(C)(C)C1=NC(C2=C(F)C(NS(C3=C(F)C=CC=C3F)(=O)=O)=CC=C2)=C(C4=CC=NC(N)=N4)S1.CS(=O)(O)=O
Molecular Weight (g/mol) 615.67
Formula Weight 615.67
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.