missing translation for 'onlineSavingsMsg'
Learn More

Dabrafenib Mesilate, Mikromol

Product Code. 30461974
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461974 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461974 Supplier Mikromol Supplier No. MM3954.000250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dabrafenib Mesilate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dabrafenib Mesylate
CAS 1195768-06-9
Analyte or Component Names Dabrafenib Mesylate
Shipping Condition Room Temperature
Molecular Formula C23 H20 F3 N5 O2 S2 . C H4 O3 S
Certifications/Compliance ISO 17034
Synonym Benzenesulfonamide, N-[3-[5-(2-amino-4-pyrimidinyl)-2-(1,1-dimethylethyl)-4-thiazolyl]-2-fluorophenyl]-2,6-difluoro-, methanesulfonate (1:1), Dabrafenib mesylate, GSK 2118436 methanesulfonate salt, GSK 2118436B, N-[3-[5-(2-Aminopyrimidin-4-yl)-2-tert-butyl-1,3-thiazol-4-yl]-2-fluorophenyl]-2,6-difluorobenzenesulfonamide methanesulfonic acid, GSK 2118436B
InChI Formula InChI=1S/C23H20F3N5O2S2.CH4O3S/c1-23(2,3)21-30-18(19(34-21)16-10-11-28-22(27)29-16)12-6-4-9-15(17(12)26)31-35(32,33)20-13(24)7-5-8-14(20)25;1-5(2,3)4/h4-11,31H,1-3H3,(H2,27,28,29);1H3,(H,2,3,4)
Solution Type Neat
SMILES CC(C)(C)c1nc(c2cccc(NS(=O)(=O)c3c(F)cccc3F)c2F)c(s1)c4ccnc(N)n4.CS(=O)(=O)O
IUPAC Name N-[3-[5-(2-aminopyrimidin-4-yl)-2-tert-butyl-1,3-thiazol-4-yl]-2-fluorophenyl]-2,6-difluorobenzenesulfonamide;methanesulfonic acid
Molecular Weight (g/mol) 615.67
Quantity 250 mg
Formula Weight 615.0892 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.