missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Dabrafenib
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
- Dabrafenib is a reversible, selective, and ATP-competitive inhibitor of B-RAF with higher potency towards V600E mutant B-RAF over wild-type B-RAF or C-RAF, with IC50 values of 0.8, 3.2 and 5 nM, respectively (Laquerre et al.; Menzies et al.)
- Decreases ERK phosphorylation and inhibits cell proliferation in cells expressing the B-RAF mutation V600E
- Prevents tumor growth in mouse xenograft models of B-RAF(V600E) human melanoma and colon cancer (Rheault et al.)
- Alleviates acetaminophen-induced liver injury through distinct nanomolar inhibition of receptor-interacting protein (RIP3) in mice (Li et al.)
Specifications
Specifications
| CAS | 1195765-45-7 |
| Synonym | GSK 2118436; Dabarefenib; Tafinlar™ |
| Quantity | 50 mg |
| Molecular Formula | C23H20F3N5O2S2 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction