missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Dabrafenib

Product Code. 17190162
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17190162 10 mg 10mg
17100172 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17190162 Supplier STEMCELL Technologies Supplier No. 73072
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MEK/ERK pathway inhibitor; Inhibits B-RAF V600E

  • Dabrafenib is a reversible, selective, and ATP-competitive inhibitor of B-RAF with higher potency towards V600E mutant B-RAF over wild-type B-RAF or C-RAF, with IC50 values of 0.8, 3.2 and 5 nM, respectively (Laquerre et al.; Menzies et al.)
  • Decreases ERK phosphorylation and inhibits cell proliferation in cells expressing the B-RAF mutation V600E
  • Prevents tumor growth in mouse xenograft models of B-RAF(V600E) human melanoma and colon cancer (Rheault et al.)
  • Alleviates acetaminophen-induced liver injury through distinct nanomolar inhibition of receptor-interacting protein (RIP3) in mice (Li et al.)

Specifications

CAS 1195765-45-7
Synonym GSK 2118436; Dabarefenib
Quantity 10 mg
Molecular Formula C23H20F3N5O2S2
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.