missing translation for 'onlineSavingsMsg'
Learn More

Dabigatran etexilate mesylate, MedChemExpress

Product Code. 16404826
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16404826

Brand: Medchem Express HY10274A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dabigatran etexilate mesylate (BIBR 1048MS) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate mesylate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.

Dabigatran etexilate mesylate (BIBR 1048MS) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate mesylate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.

Specifications

Chemical Name or Material Dabigatran etexilate mesylate
CAS 872728-81-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C35H45N7O8S
Quantity 100 mg
Synonym BIBR 1048MS Dabigatran etexilate methanesulfonate
Solubility Information DMSO : 50 mg/mL (69.08 mM; Need ultrasonic)
SMILES O=C(N(C1=NC=CC=C1)CCC(OCC)=O)C2=CC=C3C(N=C(N3C)CNC4=CC=C(C(NC(OCCCCCC)=O)=N)C=C4)=C2.CS(=O)(O)=O
Molecular Weight (g/mol) 723.84
Formula Weight 723.84
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.