missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAB2IP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23582-1-AP-150UL
This item is not returnable.
View return policy
Description
DAB2IP is a Ras GTPase-activating protein (GAP) that acts as a tumor suppressor. The DAB2IP gene is inactivated by methylation in prostate and breast cancers.Specifications
| DAB2IP | |
| Polyclonal | |
| Unconjugated | |
| DAB2IP | |
| AF9Q34, AIP1, ASK interacting protein 1, DAB2 interacting protein, DAB2 interaction protein, DAB2IP, DIP1/2, KIAA1743 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 153090, 69601 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.72 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3UHC7, Q5VWQ8 | |
| Dab2ip | |
| DAB2IP Fusion Protein Ag20308 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction