missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAB2IP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585697
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (98%), Rat (98%), Bovine (98%). Positive Control: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DAB2IP is a Ras GTPase-activating protein (GAP) that acts as a tumor suppressor. The DAB2IP gene is inactivated by methylation in prostate and breast cancers.
Specifications
| DAB2IP | |
| Polyclonal | |
| Unconjugated | |
| Dab2ip | |
| 2310011D08Rik; AF9Q34; AI480459; AIP1; AIP-1; ASK1-interacting protein 1; ASK-interacting protein 1; DAB2 interacting protein; DAB2 interaction protein; DAB2-interacting protein; Dab2ip; DIP1/2; disabled 2 interacting protein; disabled 2 interacting protein long form; disabled homolog 2 interacting protein; disabled homolog 2-interacting protein; DOC2/DAB2 interactive protein; DOC-2/DAB2 interactive protein; DOC-2/DAB-2 interactive protein; Kiaa1743; mKIAA1743; nGAP-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 153090 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.06 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q5VWQ8 | |
| Dab2ip | |
| Recombinant protein encompassing a sequence within the C-terminus region of human DAB2IP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction