missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DAB2 Recombinant Rabbit Monoclonal Antibody (K01_2M07)

Product Code. 30826401
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826401 100 μL
30828789 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826401 Supplier Invitrogen™ Supplier No. MA564040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

DAB2 Recombinant Monoclonal Antibody for IHC (P), Western Blot

DAB2 mRNA is expressed in normal ovarian epithelial cells but is down-regulated or absent from ovarian carcinoma cell lines. The 770-amino acid predicted protein has an overall 83% identity with the mouse p96 protein, a putative mitogen-responsive phosphoprotein; homology is strongest in the amino-terminal end of the protein in a region corresponding to the phosphotyrosine interaction domain. The down-regulation of DAB2 may play an important role in ovarian carcinogenesis. This gene was initially named DOC2 (for Differentially expressed in Ovarian Cancer) and is distinct from the DOC2A and DOC2B genes (for double C2-like domains, alpha and beta).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAB2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2M07
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene DAB2
Gene Accession No. P98082
Gene Alias 5730435J12Rik; AA960054; Adaptor molecule disabled-2; AI957090; C9; D15Wsu122e; D630005B22Rik; Dab, mitogen-responsive phosphoprotein, homolog 2; Dab2; DAB2, clathrin adaptor protein; differentially expressed in ovarian carcinoma 2; Differentially-expressed protein 2; disabled 2, mitogen-responsive phosphoprotein; disabled homolog 2; disabled homolog 2 mitogen-responsive phosphoprotein; disabled homolog 2, mitogen-responsive phosphoprotein; Doc2; Doc-2; DOC-2 p82 isoform; FLJ26626; Mitogen-responsive phosphoprotein; p96
Gene Symbols DAB2
Host Species Rabbit
Immunogen A synthetic peptide of human DAB2.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1601
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.