missing translation for 'onlineSavingsMsg'
Learn More

DAB2 Antibody [PE], Novus Biologicals™

Product Code. 30737315 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737315 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737315 Supplier Novus Biologicals Supplier No. AF8064P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

DAB2 Polyclonal antibody specifically detects DAB2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAB2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias Differentially-expressed protein 2, disabled (Drosophila) homolog 2 (mitogen-responsive phosphoprotein), disabled homolog 2, mitogen-responsive phosphoprotein (Drosophila), DOC-2DOC2disabled homolog 2, FLJ26626, mitogen-responsive phosphoprotein
Host Species Goat
Immunogen E. coli-derived recombinant human DAB2, Lys630-Ala770, Accession # P98082
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1601
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.