missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ DAB Advanced Chromogenic Kit

Product Code. 15521247
Change view
Click to view available options
Quantity:
50 tests
Unit Size:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15521247 50 tests 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Reagent A (DAB Chromogen) is a suspected carcinogen, avoid contact with skin and eyes and dispose in accordance with local regulations. Reagent B (DAB Diluent) contains hydrogen peroxide solution, avoid contact with skin and eyes.
Product Code. 15521247 Supplier Invitrogen™ Supplier No. 8801496572
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Reagent A (DAB Chromogen) is a suspected carcinogen, avoid contact with skin and eyes and dispose in accordance with local regulations. Reagent B (DAB Diluent) contains hydrogen peroxide solution, avoid contact with skin and eyes.

Includes

  • Reagent A (DAB Chromogen)
  • Reagent B (DAB Diluent)
  • Labeled Empty Dropper Bottle : for storage and application of working DAB solution

Contains the necessary reagents to prepare a DAB substrate working solution.

  • The enzyme peroxidase uses the DAB substrate to produce a brownish-red precipitate
  • These precipiates are insoluble in organic solvent
  • This enzyme/substrate reaction can be used on tissue sections or cells (grown on culture slides or cytospins) to visualize the localization of an HRP-conjugated antibody
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Colorimetric
Quantity 50 tests
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase) Substrate
Product Line eBioscience
Product Type DAB Advanced Chromogenic Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.