missing translation for 'onlineSavingsMsg'
Learn More

DAAO inhibitor-1, MedChemExpress

Product Code. 16497167
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497167 5 mg 5mg
16490757 50 mg 50mg
16454448 10 mg 10mg
16405336 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497167 Supplier Medchem Express Supplier No. HY111412
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress DAAO inhibitor-1 is a potent D-amino acid oxidase (DAAO) inhibitor with an IC50 of 0.12 μM.

DAAO inhibitor-1 is a potent D-amino acid oxidase (DAAO) inhibitor with an IC50 of 0.12 μM.

Specifications

Chemical Name or Material DAAO inhibitor-1
CAS 2065250-25-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C7H5FN2O
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (821.67 mM; Need ultrasonic)
SMILES FC1=CC=C2C(NN=C2O)=C1
Molecular Weight (g/mol) 152.13
Formula Weight 152.13
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.