Learn More
DAAM1 Mouse anti-Human, Unconjugated, Clone: 3F11F11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67287-1-IG-20UL
This item is not returnable.
View return policy
Description
The DAAM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DAAM1. This antibody recognizes human, mouse, rat antigen. The DAAM1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| DAAM1 | |
| Monoclonal | |
| 920 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 3F11F11 | |
| Unconjugated | |
| 20 μL | |
| 23002 | |
| IgG1 |