missing translation for 'onlineSavingsMsg'
Learn More

DAAM1 Antibody (4H3), Novus Biologicals™

Product Code. 18320419 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320419 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320419 Supplier Novus Biologicals Supplier No. H00023002M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DAAM1 Monoclonal antibody specifically detects DAAM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Proximity Ligation Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAAM1
Applications Western Blot, ELISA, Immunoprecipitation, Proximity Ligation Assay
Classification Monoclonal
Clone 4H3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunoprecipitation, Proximity Ligation Assay
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055807
Gene Alias disheveled-associated activator of morphogenesis 1, dishevelled associated activator of morphogenesis 1, dishevelled-associated activator of morphogenesis 1, KIAA0666FLJ41657
Host Species Mouse
Immunogen DAAM1 (NP_055807, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAPRKRGGRGISFIFCCFRNNDHPEITYRLRNDSNFALQTMEPALPMPPVEELDVMFSELVDELDLTDKHREAMFALPAEKKWQIYCSKKKDQEENKGATSWPEFYIDQL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 23002
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.