missing translation for 'onlineSavingsMsg'
Learn More

DAA-1106, MedChemExpress

Product Code. 16480438
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480438 5 mg 5mg
16463716 25 mg 25mg
16447978 10 mg 10mg
16423907 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16480438 Supplier Medchem Express Supplier No. HY19945

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DAA1106 is a potent and selective ligand for peripheral benzodiazepine receptor (PBR), as a potent and selective agonist at the peripheral benzodiazepine receptor.

DAA1106 is a potent and selective ligand for peripheral benzodiazepine receptor (PBR), as a potent and selective agonist at the peripheral benzodiazepine receptor.

Specifications

Chemical Name or Material DAA-1106
CAS 220551-92-8
Color White
Purity Grade Notes Research
Molecular Formula C23H22FNO4
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (252.90 mM; Need ultrasonic)
SMILES CC(N(CC1=CC(OC)=CC=C1OC)C2=CC(F)=CC=C2OC3=CC=CC=C3)=O
Molecular Weight (g/mol) 395.42
Formula Weight 395.42
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.