missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DA ZP1
GREENER_CHOICE

Product Code. 18725825
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
This item is not returnable. View return policy

Product Code. 18725825

Brand: Tocris Bioscience™ 7444/2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorogenic Zn(II) sensor for isolation of stem cell derived β cells

DA ZP1 (diacetylated Zinpyr1) is a fluorogenic Zn(II) sensor (Kd Zn2+ = 0.6 nM). It can be used to image and isolate pancreatic β-cells and β-like cells in vitro. Used to purify live stem cell-derived β-like cells. It can image transplanted islet grafts and endogenous mouse islets in vivo. DA ZP1 is non-fluorescent in the absence of Zn(II) ions but its binding with Zn(II) promotes hydrolytic cleavage of the acetyl groups, generating a strong fluorescent signal. Excitation and emission maxima (λ) are 490 nm and 522 nm, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3 ',6 '-bis(Acetyloxy)-4 ',5 '-bis[[bis(2-pyridinylmethyl)amino]methyl]-2 ',7 '-dichlorospiro[isobenzofuran-1(3 H),9 '-[9 H]xanthen]-3-one
CAS 1816254-69-9
Quantity 2 mg
Target Fluorescent Cell Indicators and Sensors
Molecular Formula C50H40Cl2N6O7
Purity 0.9
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.