Learn More
3M™ Secure Click™ Particulate Filter P1 R, D7915
Product Code. 16416390
Change view
Click to view available options
Filter Type:
P1R
Model:
D7915
Unit Size:
10 pairs
Product Code. Filter Type Model unitSize
16416390 P1R D7915 10 pairs
1 options
Product Code. 16416390 Supplier 3M™ Supplier No. 7100197283
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Secure Click™ Particulate Filter P1 R D7915 is CE approved for environments containing certain solid and liquid particles. Use our particulate filter with 3M™ Secure Click™ Respirators 800 Series, 3M™ Secure Click™ filters D8000 Series and 3M™ Filter Retainer D701.
  • New dual flow cartridges and filters allow for easier breathing
  • Protects against particulates
  • Quick, simple connection. Simply align the connections and push until you hear a click
  • Use with 3M™ Secure Click™ Reusable Respirators 800 Series
  • Swept-back design allows an enhanced field of view and comfort
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved
Connection Type Secure Click™
Filter Type P1R
Product Type Replacement Filter
For Use With (Equipment) 3M™ Secure Click™ Respirators 800 Series, Secure Click™ Filter Cartridges D8000 Series, and Filter Retainer D701
Model D7915