missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D4S234E Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-26148
This item is not returnable.
View return policy
Description
D4S234E Polyclonal specifically detects D4S234E in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| D4S234E | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| carboxyterminally EE-tagged neuron-enriched endosomal 21 kDa protein, D4S234Brain neuron cytoplasmic protein 1, DNA segment on chromosome 4 (unique) 234 expressed sequence, NEEP21, neuron-specific protein family member 1, NSG1, P21 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 5 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| NP_077042.2 | |
| NSG1 | |
| Peptide with sequence EKGTKQPLLEDGFD-C corresponding to N-Terminus according to NP_077042.2. | |
| 0.1 mg | |
| Signal Transduction | |
| 27065 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction