missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies D4476

Product Code. 17100312
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 17100312

Brand: STEMCELL Technologies 74012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Activin/NODAL/TGF-β pathway inhibitor; Inhibits CK1, ALK5 and p38 MAPK

  • D4476 is a selective, potent, and cell-permeant inhibitor of casein kinase (CK) 1 (Rena et al.) It inhibits CK1 and CK1δ from S
  • pombe (IC50 = 200 nM and 300 nM, respectively; Bain et al.; Rena et al.) It also inhibits transforming growth factor β1 (TGF-β1) type I receptor (ALK5) with IC50 = 500 nM (Rena et al.)
  • Along with forskolin (Catalog #72112) and 2-methyl-5-hydroxytryptamine, reprograms mouse somatic cells into pluripotent cells by acting as a chemical substitute for OCT4 (Hou et al.)
  • Reprograms mouse somatic cells into pluripotent cells by acting as a chemical substitute for OCT4 (Hou et al.)
  • Induces apoptosis of multiple myeloma cell lines (Hu et al.)
  • Exhibits anti-leukemic activity by selectively killing leukemia stem cells over normal hematopoietic stem and progenitor cells (Järås et al.)

Specifications

CAS 301836-43-1
Synonym Casein Kinase I Inhibitor
Quantity 5 mg
Molecular Formula C23H18N4O3
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.